Please login to your online account to display your discounted pricing

Trace Metals 2 For Environmental Analysis, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 500mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  WP3500

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. WP-3-500

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Concentration or Composition (by Analyte or Components) Components in 2% HNO3:
Sb    •20μg/mL
Ag    •10μg/mL
Tl    •20μg/mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Packaging Triple-leached LDPE bottle
Chemical Name or Material Trace Metals 2
Solvent or Matrix 2% HNO3
DOT Information Hazardous
Recommended Storage Ambient
Purity Grade For Environmental Analysis
Name Note Groundwater and Wastewater Pollution Control Check Standard
CAS 7697-37-2

Specifications

Chemical Name or Material Trace Metals 2
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sb20μg/mL
Tl20μg/mL
Ag10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Ambient