missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trace Metals 1 without Mercury For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  Spex Certiprep Llc WP15N

 View more versions of this product

Catalog No. WP15N


 

Disclaimers:

Product intended for laboratory use only


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Specifications

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al500μg/mL
V250μg/mL
As100μg/mL
Be100μg/mL
Co100μg/mL
Cr100μg/mL
Cu100μg/mL
Fe100μg/mL
Mn100μg/mL
Ni100μg/mL
Pb100μg/mL
Zn100μg/mL
Cd25μg/mL
Se25μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Description & Specifications

Specifications

Trace Metals I without Mercury
Various
Various
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
125mL
Groundwater and Wastewater Pollution Control Check Standard
For Environmental Analysis
Triple-leached LDPE bottle
5% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Chemical Identifiers

AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Trace Metals I without Mercury

Safety and Handling

Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.

DOTInformation : Hazardous

Recommended Storage : Ambient