missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TRA-1-81 (podocalyxin) Mouse anti-Human, PE, Clone: TRA-1-81, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $302.28

Specifications

Antigen TRA-1-81 (podocalyxin)
Clone TRA-1-81
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011227 Life Technologies
LS12888380
25μg Each for $130.68
5011228 Life Technologies
LS12888382
100μg Each for $302.28
Specifications

Description

The TRA-1-81 antibody recognizes a protein expressed on undifferentiated human embryonic stem cells (ES), embyronal carcinoma cells (EC), and embryonic germ cells (EG). Like other stem cell specific markers, the epitope recognized by the TRA-1-81 antibody is lost upon cell differentiation. The TRA-1-81 epitope is resistant to neuraminidase digestion, unlike the epitope recognized by the related TRA-1-60 antibody. The TRA-1-81 antibody is known to specifically recognize a carbohydrate epitope on a keratan sulfated glycoprotein recently identified as podocalyxin, a member of the CD34-related family of sialomucins. Podocalyxin is a transmembrane glycoprotein originally identified on epithelial glomerular cells known as podocytes, and the protein has also been implicated in the development of aggressiveness in a variety of cancers, including breast and prostate cancer.
Description & Specifications

Specifications

TRA-1-81 (podocalyxin)
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TRA-1-81
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL