Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ TR2/HVEM Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ TR2/HVEM Ab-1, Mouse Monoclonal Antibody.

$510.65 - $1,683.22

Specifications

Antigen TR2/HVEM Ab-1
Clone 122
Applications Flow Cytometry
Applications Immunofluorescence
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS710P

 
thermo scientific™ lab vision™
MS710P
200μg/mL 1mL Each for $1,556.14

MS710P0

 
thermo scientific™ lab vision™
MS710P0
200μg/mL 100μL Each for $510.65

MS710P1

 
thermo scientific™ lab vision™
MS710P1
200μg/mL 500μL Each for $1,079.02

MS710P1ABX

 
thermo scientific™ lab vision™
MS710P1ABX
1mg/mL 100μL Each for $1,191.14

MS710PABX

 
thermo scientific™ lab vision™
MS710PABX
1mg/mL 200μL Each for $1,683.22
Description & Specifications

Specifications

Antigen TR2/HVEM Ab-1
Clone 122
Applications Flow Cytometry
Applications Immunofluorescence
Applications Flow Cytometry
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Infectious Disease
Type Primary
Immunogen Recombinant human TR2 protein
Isotype IgG1

Provide accurate, reproducible results with the Thermo Scientific™ TR2/HVEM Ab-1, Mouse Monoclonal Antibody.

TNF receptor like-2 (TR2) (also known as herpesvirus entry mediator (HVEM)) is expressed in multiple human tissues and cell lines and shows a constitutive and relatively high expression in peripheral T and B lymphocytes, and monocytes. It is a member of TNF receptor family. TR2/HVEM is a receptor for herpes virus. It also acts as a receptor for LIGHT (also known as HVEM ligand), a member of TNF family. TR2 protein inhibits a mixed lymphocyte reaction-mediated proliferation suggesting that the receptor and/or its ligand play a role in T cell stimulation.

Host Species: Mouse

Clone: 122

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human TR2 protein

Molecular Weight: 32kDa

Positive Control: Tonsil

Cellular Localization: Secreted protein

Recommended for:

  • Immunofluorescence
  • Flow Cytometry
  • Function Blocking

Note:

USA: RUO; Int'l: RUO