Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ TR2/HVEM Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ TR2/HVEM Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS710P1ABX

 View more versions of this product

Catalog No. MS710P1ABX

  • / Each
 

Description & Specifications

Specifications

Isotype IgG1
Cross Reactivity Human
Regulatory Status RUO
Antigen TR2/HVEM Ab-1
Host Species Mouse
Applications Flow Cytometry
Applications Immunofluorescence
Format Concentrated
Immunogen Recombinant human TR2 protein
Quantity 100µL
Clone 122
Concentration 1mg/mL
Conjugate Unlabeled
Research Discipline Infectious Disease
Type Primary
Classification Monoclonal

Provide accurate, reproducible results with the Thermo Scientific™ TR2/HVEM Ab-1, Mouse Monoclonal Antibody.

TNF receptor like-2 (TR2) (also known as herpesvirus entry mediator (HVEM)) is expressed in multiple human tissues and cell lines and shows a constitutive and relatively high expression in peripheral T and B lymphocytes, and monocytes. It is a member of TNF receptor family. TR2/HVEM is a receptor for herpes virus. It also acts as a receptor for LIGHT (also known as HVEM ligand), a member of TNF family. TR2 protein inhibits a mixed lymphocyte reaction-mediated proliferation suggesting that the receptor and/or its ligand play a role in T cell stimulation.

Host Species: Mouse

Clone: 122

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human TR2 protein

Molecular Weight: 32kDa

Positive Control: Tonsil

Cellular Localization: Secreted protein

Recommended for:

  • Immunofluorescence
  • Flow Cytometry
  • Function Blocking

Note:

USA: RUO; Int'l: RUO