Please login to your online account to display your discounted pricing

Tr. Met. Standards (Set 3 Grndwtr. Wstwtr. Pollut. Ctrl. Chk. Standards/For Env. Anal.), SPEX CertiPrep™

$1,267.49 - $1,267.49

Chemical Identifiers

Chemical Name or Material Trace Metals
Name Note Set of 3 Groundwater and Wastewater Pollution Control Check Standard
Purity Grade For Environmental Analysis
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TM-SET

 
spex certiprep llc
TMSET
Triple-leached LDPE bottle (3) each 125mL Each for $1,267.49
Description & Specifications

Specifications

Quantity (3) each 125mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Health Hazard 1 Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Hazardous
Chemical Name or Material Trace Metals
Packaging Triple-leached LDPE bottle
Purity Grade For Environmental Analysis
Name Note Set of 3 Groundwater and Wastewater Pollution Control Check Standard

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.

Ambient