Please login to your online account to display your discounted pricing

TQS, Tocris™

Positive allosteric modulator of α7 nAChRs

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material TQS
Assay Percent Range >98%
Molecular Formula C22H20N2O2S
Formula Weight 376.47
Synonym 3a,4,5,9b-Tetrahydro-4-(1-naphthalenyl)-3H-cyclopentan[c]quinoline-8-sulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

423310

 
r & d systems
423310
10mg Each for $225.77

423350

 
r & d systems
423350
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 376.47
Quantity 10mg
Chemical Name or Material TQS
Assay Percent Range >98%
Molecular Formula C22H20N2O2S
CAS 353483-92-8
Synonym 3a,4,5,9b-Tetrahydro-4-(1-naphthalenyl)-3H-cyclopentan[c]quinoline-8-sulfonamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C