Epredia™ TPX Single Sample Chamber, Caps and Filter Cards
For use with disposable white filter cards for sample volumes up to 0.5 mL and for use with disposable brown filter cards for sample volumes of 0.4 mL or less.
$168.03 - $4,506.03
Products 3
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
5991022 Edge
View Documents Promotion Details
Epredia™
5991022
White Filter Cards
Case of 200 Each for $168.03
Edge
28-618-351
View Documents Promotion Details
Epredia™
A78710018
TPX Sample Chamber
Case of 12 Each for $4,506.03
5991023
View Documents Promotion Details
Epredia™
5991023
Brown Filter Cards
Case of 200 Each for $207.23
Description
Description
TPX Single Sample Chamber
  • Reusable
  • Autoclavable
Filter Cards
  • Highly absorbent disposable filter cards for use with TPX chamber
  • Not for use with Cytofunnel™ disposable sample chambers