Please login to your online account to display your discounted pricing

TPMPA, Tocris


Selective GABAC antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS104010

 View more versions of this product

Catalog No. 104010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 161.14
Quantity 10mg
Chemical Name or Material TPMPA
Assay Percent Range >98%
CAS 182485-36-5
Recommended Storage Store at Room Temperature
Molecular Formula C6H12NO2P
Synonym (1,2,5,6-Tetrahydropyridin-4-yl)methylphosphinic acid

Specifications

Chemical Name or Material TPMPA
Assay Percent Range >98%
Molecular Formula C6H12NO2P
Formula Weight 161.14
Synonym (1,2,5,6-Tetrahydropyridin-4-yl)methylphosphinic acid
Accessories

Store at Room Temperature