Please login to your online account to display your discounted pricing

TP 003, Tocris™

Functionally selective GABAA receptor (α3 subtype) partial agonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material TP 003
Assay Percent Range >98%
Molecular Formula C23H16F3N3O
Formula Weight 407.39
Synonym 2',4-Difluoro-5'-[8-fluoro-7-(1-hydroxy-1-methylethyl)imidazo[1,2-a]-pyridin-3-yl]-[1,1'-biphenyl]-2-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

441410

 
r & d systems
441410
10mg Each for $239.63

441450

 
r & d systems
441450
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 407.39
Quantity 10mg
Chemical Name or Material TP 003
Molecular Formula C23H16F3N3O
Assay Percent Range >98%
Synonym 2',4-Difluoro-5'-[8-fluoro-7-(1-hydroxy-1-methylethyl)imidazo[1,2-a]-pyridin-3-yl]-[1,1'-biphenyl]-2-carbonitrile
CAS 628690-75-5
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C