Please login to your online account to display your discounted pricing

Toxaphene (For EPA 500, 600, 8000/; Appendix IX, CLP Methods)


Toxaphene at 100μg/mL in methanol or hexane

Quantity 1x1mL (in Methanol)
Packaging Ampule

Manufacturer: ultra scientific  PP2701

 View more versions of this product

Catalog No. US-PP-2701

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL (in Methanol)
Methods For EPA 500, 600, 8000/Appendix IX
Chemical Name or Material Toxaphene
CAS 8001-35-2
Packaging Ampule

Specifications

Chemical Name or Material Toxaphene
CAS 8001-35-2