missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ansell™ TouchNTuff™ Disposable Chemical Resistant Nitrile Gloves

Proven splash resistance against hazardous chemicals.

Manufacturer:  Ansell™ 92605S114096

 View more versions of this product

Catalog No. 19-148-540




Description

Description

  • Contains no natural rubber latex proteins, which prevents from Type I allergies for the wearer
  • Strong grip in wet or dry conditions
  • Rolled beaded for easy donning
  • Powder-Free, Polymer-coated 
  • Ambidextrous 
  • Textured Fingers
  • Length: 300mm (11.8 in.)
  • Palm Thickness : 0.12mm (4.7 mil)
  • Color: Green
  • AQL: 1.5

Compliance

EN420, EN374/1,2,3, ASTM D 6319, FDA 21 CFR 177-2600

Specifications

Specifications

Yes
Beaded/Rolled
Laboratory and utility
Nitrile
92-605
>14 Mpa
0.12mm
Powder-free
11.8 in.
Textured
Ambidextrous
Yes
Hazardous chemicals
Small
4.7 mil
Chemical-resistant glove
Green
300mm
Documents