Please login to your online account to display your discounted pricing

Total Nitric Oxide and Nitrate/Nitrite Parameter ELISA Kit, R&D Systems

$1,953.58 - $1,953.58

Specifications

Cross Reactivity < 50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Quantity 1 Pk.
For Use With (Application) ELISA
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

SKGE001

 
r & d systems
RDSSKGE001
ELISA kit Each for $1,953.58
Description & Specifications

Specifications

Cross Reactivity < 50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Quantity 1 Pk.
For Use With (Application) ELISA

Assay Type: Solid Phase Competitive ELISA
Sample Type/Volume Required: Cell Culture Supernates (10μL), Serum (25μL), EDTA Plasma (25μL), Heparin Plasma (25μL), Citrate Plasma (25μL), Urine (10μL)
Sensitivity: 0.78 μmol/L
Specificity: Measures Total Nitric Oxide, Nitrite and Nitrate levels in various samples
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.