missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Total MMP-2 Quantikine ELISA Kit, R&D Systems

a 4.5 hour solid-phase ELISA designed to measure MMP-2 in cell culture supernates, serum, and plasma.

$624.40 - $2,985.15

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Species Human, Mouse, Rat, Porcine, Canine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MMP200 R & D Systems
RDSMMP200
1 Plate Each for $624.40
SMMP200 R & D Systems
RDSSMMP200
6 Plates Each for $2,985.15
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Sensitivity: 0.082ng/mL
Specificity: Recombinant MMP-2 and natural human, mouse, rat, porcine, and canine active, pro-, and TIMP complexed MMP-2.
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Solid Phase Sandwich ELISA
Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Human, Mouse, Rat, Porcine, Canine