Please login to your online account to display your discounted pricing

Torin 2, Tocris™

Potent and selective mTOR inhibitor

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material Torin 2
Assay Percent Range >98%
Molecular Formula C24H15F3N4O
Formula Weight 432.4
Synonym 9-(6-Amino-3-pyridinyl)-1-[3-(trifluoromethyl)phenyl]-benzo[h]-1, 6-naphthyridin-2(1H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

424810

 
r & d systems
RDS424810
10mg Each for $284.81

424850

 
r & d systems
RDS424850
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 432.4
Quantity 10mg
Assay Percent Range >98%
Synonym 9-(6-Amino-3-pyridinyl)-1-[3-(trifluoromethyl)phenyl]-benzo[h]-1, 6-naphthyridin-2(1H)-one
Chemical Name or Material Torin 2
Molecular Formula C24H15F3N4O
CAS 1223001-51-1
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C