Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Topoisomerase II α Ab-4, Mouse Monoclonal Antibody

Topoisomerase II α Ab-4, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

$288.98 - $1,632.92

Specifications

Antigen Topoisomerase II alpha Ab-4
Clone Ki-S1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1819R7

 
thermo scientific™ lab vision™
MS1819R7
7mL Each for $625.82

MS1819S

 
thermo scientific™ lab vision™
MS1819S
1mL Each for $1,632.92

MS1819S0

 
thermo scientific™ lab vision™
MS1819S0
100μL Each for $288.98

MS1819S1

 
thermo scientific™ lab vision™
MS1819S1
500μL Each for $954.06
Description & Specifications

Specifications

Antigen Topoisomerase II alpha Ab-4
Clone Ki-S1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Murine
Regulatory Status IVD
Research Discipline DNA Repair
Type Primary
Immunogen Lysate of the human histiocytic leukemia cell line U937

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ Topoisomerase II α Ab-4, Mouse Monoclonal Antibody.

Topo II α plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2- & M-phases in transformed and developmentally regulated normal cellsTopo II α is also implicated in drug resistance of tumor cells.

Host Species: Mouse

Clone: Ki-S1

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Lysate of the human histiocytic leukemia cell line U937

Molecular Weight: 170kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)