Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Topoisomerase II α Ab-1, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Topoisomerase II α Ab-1, Rabbit Polyclonal Antibody.

$275.39 - $1,223.27

Specifications

Antigen Topoisomerase II alpha Ab-1
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB117P

 
thermo scientific™ lab vision™
RB117P
Purified Ab with BSA and Azide 1mL Each for $1,125.84

RB117P0

 
thermo scientific™ lab vision™
RB117P0
Purified Ab with BSA and Azide 100µL Each for $275.39

RB117P1

 
thermo scientific™ lab vision™
RB117P1
Purified Ab with BSA and Azide 500µL Each for $739.10

RB117P1ABX

 
thermo scientific™ lab vision™
RB117P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $836.55

RB117PABX

 
thermo scientific™ lab vision™
RB117PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,223.27
Description & Specifications

Specifications

Antigen Topoisomerase II alpha Ab-1
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen A synthetic peptide, corresponding to aa 1513-1530 (RAKKPIKYLEESDEDDLF) from the C-terminus of human topoisomerase IIalpha
Isotype IgG

Provide accurate, reproducible results with the Thermo Scientific™ Topoisomerase II α Ab-1, Rabbit Polyclonal Antibody.

Topo IIα plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2- & M-phases in transformed and developmentally regulated normal cells. Note that Ki67/M1B1 is also present in G1-phase. Topo IIα is also implicated in drug resistance of tumor cells.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: aa 1513-1530

Immunogen: A synthetic peptide, corresponding to aa 1513-1530 (RAKKPIKYLEESDEDDLF) from the C-terminus of human topoisomerase IIα

Molecular Weight: 170kDa

Positive Control: Raji cells or any exponentially growing cultured human cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO