Please login to your online account to display your discounted pricing

Tomoxetine hydrochloride, Tocris

Potent, selective noradrenalin re-uptake inhibitor

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material Tomoxetine hydrochloride
Assay Percent Range >99%
Molecular Formula C17H21NO·HCl
Formula Weight 291.82
Synonym (R)-N-Methyl-γ-(2-methylphenoxy)-benzenepropanamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

201110

 
r & d systems
RDS201110
10mg Each for $214.12

201150

 
r & d systems
RDS201150
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 291.82
Quantity 10mg
Assay Percent Range >99%
Chemical Name or Material Tomoxetine hydrochloride
Molecular Formula C17H21NO·HCl
CAS 82248-59-7
Synonym (R)-N-Methyl-γ-(2-methylphenoxy)-benzenepropanamine hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature