Learn More
Tolylene 2,4-diisocyanate, 80%, tech., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 202502500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tolylene 2,4-diisocyanate | |
12.5°C to 13.5°C | |
1.2200g/mL | |
127°C | |
75% min. (GC) | |
C9H6N2O2 | |
CH3C6H3(NCO)2 | |
13,IV,243 | |
15,9689 | |
Solubility in water: reacts with water. Other solubilities: evolving carbon dioxide in water, soluble in ether, acetone and other organic solven | |
CC1=C(C=C(C=C1)N=C=O)N=C=O | |
174.16 | |
CHEBI:53556 | |
80% |
584-84-9 | |
Colorless to Yellow | |
115.0°C to 120.0°C (10.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5670 to 1.5690 | |
250 mL | |
1.22 | |
2,4-toluene diisocyanate, 2,4-diisocyanatotoluene, tolylene-2,4-diisocyanate, toluene-2,4-diisocyanate, 2,4-tolylene diisocyanate, toluene 2,4-diisocyanate, 4-methyl-m-phenylene diisocyanate, tolylene diisocyanate, 2,4-tdi, toluylene-2,4-diisocyanate | |
DVKJHBMWWAPEIU-UHFFFAOYSA-N | |
2,4-diisocyanato-1-methylbenzene | |
11443 | |
174.16 | |
Liquid |
Chemical Identifiers
584-84-9 | |
174.16 | |
2,4-toluene diisocyanate, 2,4-diisocyanatotoluene, tolylene-2,4-diisocyanate, toluene-2,4-diisocyanate, 2,4-tolylene diisocyanate, toluene 2,4-diisocyanate, 4-methyl-m-phenylene diisocyanate, tolylene diisocyanate, 2,4-tdi, toluylene-2,4-diisocyanate | |
CHEBI:53556 | |
CC1=C(C=C(C=C1)N=C=O)N=C=O |
C9H6N2O2 | |
DVKJHBMWWAPEIU-UHFFFAOYSA-N | |
11443 | |
2,4-diisocyanato-1-methylbenzene |
Safety and Handling
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Fatal if inhaled.
May cause an allergic skin reaction.
Suspected of cau
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritat
GHS Signal Word: Danger
EINECSNumber : 209-544-5
RUO – Research Use Only