Learn More
Benzyl mercaptan, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 421261000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzyl mercaptan | |
98.5 | |
-29°C | |
1.0580g/mL | |
70°C | |
98.5% min. (GC) | |
C7H8S | |
C6H5CH2SH | |
100 g | |
1.058 | |
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan | |
UENWRTRMUIOCKN-UHFFFAOYSA-N | |
phenylmethanethiol | |
7509 | |
99% |
100-53-8 | |
100.0 | |
Colorless to Yellow | |
195°C | |
Authentic | |
Glass bottle | |
1.5735 to 1.5775 | |
MFCD00004867,MFCD00801588,MFCD01863867 | |
06, IV, 2632 | |
15, 9475 | |
Solubility in water: insoluble | |
SCC1=CC=CC=C1 | |
124.20 | |
124.21 | |
Liquid |
Chemical Identifiers
100-53-8 | |
124.20 | |
UENWRTRMUIOCKN-UHFFFAOYSA-N | |
7509 | |
SCC1=CC=CC=C1 |
C7H8S | |
MFCD00004867,MFCD00801588,MFCD01863867 | |
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan | |
phenylmethanethiol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth.
Do NO
GHS Signal Word: Danger
EINECSNumber : 202-862-5
Recommended Storage : Product darkens during storage
RUO – Research Use Only