missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Toluenesulfonyl chloride, 98%, ACROS Organics™

Manufacturer:  Acros Organics 170490250

 View more versions of this product

Catalog No. AC170490250


 

Specifications

Specifications

α-Toluenesulfonyl chloride
1939-99-7
100.0
≥97.5% (ex Cl)
C7H7ClO2S
MFCD00007455
11, 116
OAHKWDDSKCRNFE-UHFFFAOYSA-N
phenylmethanesulfonyl chloride
74740
Crystalline Powder
90°C to 94°C
98%
97.5
Authentic
98%
C6H5CH2SO2Cl
Glass bottle
Phenylmethanesulfonyl chloride
C1=CC=C(C=C1)CS(=O)(=O)Cl
190.641
190.64
White
25g

Chemical Identifiers

1939-99-7
190.641
OAHKWDDSKCRNFE-UHFFFAOYSA-N
74740
C7H7ClO2S
MFCD00007455
Phenylmethanesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

EINECSNumber : 217-717-1

TSCA : TSCA

SDS Documents