missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene UltimAR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 108-88-3
Molecular Formula C6H5CH3
Formula Weight 92.14
Chemical Name or Material Toluene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MKV5601 Avantor Performance
V56006
Glass 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MKV5604 Avantor Performance
V56010
Glass 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

108-88-3
C6H5CH3
92.14
Toluene

Specifications

108-88-3
92.14
1L
C6H5CH3
Glass
Toluene