Please login to your online account to display your discounted pricing

Toluene UltimAR, Macron Fine Chemicals™


macron

Quantity 1L
Packaging Glass

Manufacturer: avantor performance  V56006

 View more versions of this product

Catalog No. MKV5601

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 92.14
CAS 108-88-3
Chemical Name or Material Toluene
Packaging Glass
Molecular Formula C6H5CH3
Quantity 1L

Specifications

Chemical Name or Material Toluene
CAS 108-88-3
Molecular Formula C6H5CH3
Formula Weight 92.14