missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Toluene)tricarbonylchromium, 98%, ACROS Organics™

Chemical Identifiers

CAS 12083-24-8
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C10H8CrO3
Formula Weight 228.17
MDL Number MFCD00010583
Chemical Name or Material (Toluene)tricarbonylchromium
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC378820010 SDS Thermo Fisher Scientific
AC378820010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC378820050 SDS Thermo Fisher Scientific
AC378820050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

12083-24-8
100.0
C10H8CrO3
MFCD00010583
97.5
98%
228.17
(Toluene)tricarbonylchromium

Specifications

12083-24-8
100.0
Authentic
98%
Glass bottle
MFCD00010583
78° to 82°C
(Toluene)tricarbonylchromium
97.5
97.5% min.
Crystalline Powder or Crystals
C10H8CrO3
228.17
Yellow to yellow-orange
1g

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger