Learn More
Toluene, Spectrophotometric Grade, 99.7+%, Thermo Scientific Chemicals
Maximum level of impurities: Evaporation residue 4ppm, H2O 0.03%
Supplier: Thermo Scientific Chemicals 019376K7
Description
Toluene is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Toluene | |
108-88-3 | |
Colorless liquid | |
111°C | |
4°C (40°F) | |
Liquid | |
>99.7% | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 |
°C to 93°C | |
Colorless | |
0.867 | |
4 L | |
Aromatic | |
1.4967 | |
C7H8 | |
635760 | |
Miscible with ethanol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water. | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
Spectrophotometric |
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Safety and Handling
Wear suitable protective clothing and gloves. If swallowed seek medical advice immediately and show this container or label. If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336-H361d-H373
DOTInformation : Hazard Class: 3, Packing Group: II
EINECSNumber : 203-625-9
RTECSNumber : XS5250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only