missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene, Spectrophotometric Grade, 99.7+%, Thermo Scientific Chemicals

Maximum level of impurities: Evaporation residue 4ppm, H2O 0.03%

Supplier:  Thermo Scientific Chemicals 019376K2

 View more versions of this product

Catalog No. AA19376K2



Description

Description

Toluene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Toluene
108-88-3
0.867
1 L
Aromatic
1.4967
MFCD00008512
635760
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
Spectrophotometric
-93°C
Colorless
111°C
4°C (40°F)
Liquid
C7H8
UN1294
14,9529
Miscible with ethanol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water.
CC1=CC=CC=C1
92.14
CHEBI:17578
≥99.7%

Chemical Identifiers

108-88-3
92.14
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H315-H336;
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336-H361d-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE

EINECSNumber : 203-625-9

RTECSNumber : XS5250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only