Please login to your online account to display your discounted pricing

Toluene, Spectrophotometric Grade, >99.7%, Alfa Aesar™

Maximum level of impurities: Evaporation residue 4ppm, H2O 0.03%

$107.88 - $370.66

Chemical Identifiers

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade Spectrophotometric grade
Physical Form Liquid
Assay Percent Range >99.7%
Molecular Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA19376K7

  SDS
alfa aesar
19376K7
4L Each for $107.88

AA19376M1

  SDS
alfa aesar
19376M1
4 x 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA19376M6

  SDS
alfa aesar
19376M6
4 x 4L Each for $370.66
Description & Specifications

Specifications

Physical Form Liquid
Purity Grade Spectrophotometric grade
Formula Weight 92.14
Color Clear colorless
Quantity 4L
DOT Information Hazard Class: 3, Packing Group: II
Molecular Formula C6H5CH3
EINECS Number 203-625-9
Appearance Colorless liquid
Impurities Maximum level of impurities: Evaporation residue 4ppm, H2O 0.03%
MDL Number MFCD00008512
Health Hazard 2 Wear suitable protective clothing and gloves. If swallowed seek medical advice immediately and show this container or label. If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.
Health Hazard 1 H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.
CAS 108-88-3
Odor Aromatic
Chemical Name or Material Toluene
Assay Percent Range >99.7%
Safety and Handling

H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.

Wear suitable protective clothing and gloves. If swallowed seek medical advice immediately and show this container or label. If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.