missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene, Semiconductor Grade, 99% min., Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Acidity 0.2meq/g, Evaporation residue 2ppm, H2O 0.03%, Substances darkened by H2SO4 P.T., Sulfur compounds (as S) 0.003%, Cl 2ppm, PO4 0.5ppm

Manufacturer:  Alfa Aesar 19399AP

 View more versions of this product

Catalog No. AA19399AP



Specifications

Specifications

Toluene
Colorless liquid
C7H8
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CC1=CC=CC=C1
92.141
CHEBI:17578
Liquid
535°C (995°F)
29 hPa (22mm Hg) at 20°C
500mL
108-88-3
99% min.
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
Semiconductor
Aromatic
Colorless

Chemical Identifiers

108-88-3
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Safety and Handling

Safety and Handling

H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.

DOTInformation : Hazard Class: 3, Packing Group: II

EINECSNumber : 203-625-9

SDS Documents