Please login to your online account to display your discounted pricing

Toluene, Semiconductor Grade, 99% min., Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Acidity 0.2meq/g, Evaporation residue 2ppm, H2O 0.03%, Substances darkened by H2SO4 P.T., Sulfur compounds (as S) 0.003%, Cl 2ppm, PO4 0.5ppm

$137.26 - $137.26

Chemical Identifiers

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade Semiconductor Grade
Physical Form Liquid
Assay Percent Range 99% min.
Molecular Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA19399AP

  SDS
alfa aesar
19399AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA19399K2

  SDS
alfa aesar
19399K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA19399K7

  SDS
alfa aesar
19399K7
4L Each for $137.26

AA19399M1

  SDS
alfa aesar
19399M1
4 x 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Toluene
CAS 108-88-3
Health Hazard 1 H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.
DOT Information Hazard Class: 3, Packing Group: II
EINECS Number 203-625-9
Purity Grade Semiconductor Grade
Appearance Colorless liquid
Physical Form Liquid
Assay Percent Range 99% min.
Molecular Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Ignition Point 535°C (995°F)
Odor Aromatic
Vapor Pressure 29 hPa (22mm Hg) at 20°C
Color Clear colorless
Quantity 500mL
Safety and Handling

H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.