missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene (Scintanalyzed™), Fisher Chemical

Manufacturer:  Thermo Scientific T313SK4

 View more versions of this product

Catalog No. T313SK-4




Specifications

Specifications

Pass Test
4L
108-88-3
50 CPM max.
Pass Test
≥99.5 %
0.9 min.
C6H5CH3
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
92.141
CHEBI:17578
Liquid
≥99.5%
0.003% max.
111°C
-95°C
Toluene
Vapor Pressure: 29mbar at 20°C
50 CPM max.
0.866g/cm³
Also meets ACS specifications
0.9 min.
C7H8
Glass Bottle
Methylbenzene, Toluol, Phenyl Methane
CC1=CC=CC=C1
0.001% max.
1140
92.14
Pass Test
Scintanalyzed™
0.03% max.
Colorless

Chemical Identifiers

108-88-3
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
Methylbenzene, Toluol, Phenyl Methane
CHEBI:17578
CC1=CC=CC=C1
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

Signal Word:DANGER!

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents