Please login to your online account to display your discounted pricing

Toluene (Scintanalyzed™), Fisher Chemical

$234.00 - $324.46

Chemical Identifiers

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade Scintanalyzed™
Purity Grade Notes Also meets ACS specifications
Assay ≥99.5%
Assay Percent Range ≥99.5 %
Molecular Formula C7H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

T313-4

  Accessories SDS
fisher scientific company
T3134
Glass Bottle 4L Each for $234.00
Case of 4 Each for $936.00

T313SK-4

  SDS
fisher scientific company
T313SK4
Glass Bottle 4L Each for $324.46
Case of 4 Each for $1,297.84
Description & Specifications

Specifications

Assay ≥99.5%
Boiling Point 111°C
Melting Point −95°C
Formula Weight 92.14
Quantity 4L
Flash Point 4°C
UN Number 1294
DOT Information DOT Class 3, : Flammable Liquid
Molecular Formula C7H8
Density 0.866g/cm³
Purity Grade Scintanalyzed™
Physical Form Liquid
Color Colorless
Absorbance Pass Test
Packaging Glass Bottle
Synonym Methylbenzene
Residue after Evaporation 0.001% max.
Assay Percent Range ≥99.5 %
Additional Information Vapor Pressure: 29mbar at 20°C
Background 3H 50 CPM max.
MDL Number MFCD00008512
ChemAlert Storage Symbol Red
Sulfur Compounds 0.003% max.
Water 0.03% max.
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Identification Pass Test
LSC Efficiency 14C 0.9 min.
LSC Efficiency 3H 0.9 min.
Substances Darkened by H2SO4 Pass Test
Health Hazard 2 DANGER!
Background 14C 50 CPM max.
Chemical Name or Material Toluene
Cleanup Symbol Solvent
Purity Grade Notes Also meets ACS specifications
CAS 108-88-3
Health Hazard 1 Signal Word:DANGER!
Linear Formula C6H5CH3

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Signal Word:DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

DANGER!