Please login to your online account to display your discounted pricing

Toluene, Low water, BakerDRY™, J.T.Baker™

Specifically manufactured for use in organometallic and oligonucleotide synthesis

$295.63 - $521.13

Chemical Identifiers

Chemical Name or Material Toluene
CAS 108-88-3
Name Note Low water
Purity Grade BakerDRY™
Molecular Formula C6H5CH3
Formula Weight 92.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004474

 
avantor performance
936410
Glass Bottle 100mL Each for $295.63

02004475

 
avantor performance
936412
Narrow Mouth Amber Glass Bottle, w/ Septum 1L Each for $521.13
Description & Specifications

Specifications

Formula Weight 92.14
Chemical Name or Material Toluene
CAS 108-88-3
Molecular Formula C6H5CH3
Name Note Low water
Packaging Glass Bottle
Specific Gravity 1L = 0.86kg
Purity Grade BakerDRY™
Quantity 100mL
Flash Point 4.00°C