Please login to your online account to display your discounted pricing

Toluene (Laboratory), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  T3234

 View more versions of this product

Catalog No. T323-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Laboratory Grade
Melting Point −95°C
Formula Weight 92.14
Boiling Point 111°C
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Molecular Formula C7H8
Density 0.866g/cm³
Vapor Pressure 29mbar at 20°C
Color Colorless
Physical Form Liquid
Viscosity 0.6 mPa/s at 20°C
Packaging Glass Bottle
Synonym Methylbenzene
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Cleanup Symbol Solvent
Health Hazard 1 Signal Word:DANGER!
MDL Number MFCD00008512
ChemAlert Storage Symbol Red
Additional Information Vapor Pressure: 29mbar at 20°C
CAS 108-88-3
Identification Pass Test
Linear Formula C6H5CH3
Chemical Name or Material Toluene

Specifications

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade Laboratory Grade
Molecular Formula C7H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Synonym Methylbenzene
Accessories

Accessories for Catalog Number: T3234

Item Mfr. No. Item Description Price Per Unit Quantity  
296355104
Spilkleen/Cep  CEP LAB SPILL KIT SOLVENTS
spilkleen/cep
CSKLSK1SOL
Each for $341.99
03-439
Eagle™ Thermoplastics™ Safety Bottle Carriers
fisher scientific
B102
Safety bottle carriers are molded-in handle Each for $217.50
Accessories
 

Signal Word:DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

DANGER!

 

SDS & Safety Documents