Please login to your online account to display your discounted pricing

Toluene (Laboratory), Fisher Chemical


Quantity 20L
Packaging Steel Pail

Manufacturer: fisher scientific company  T32320

 View more versions of this product

Catalog No. T323-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Laboratory Grade
Boiling Point 111°C
Melting Point −95°C
Formula Weight 92.14
Quantity 20L
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 29mbar at 20°C
Vapor Density 3.1g/mol
Viscosity 0.6 mPa/s at 20°C
Physical Form Liquid
Color Colorless
Molecular Formula C7H8
Density 0.866g/cm³
Identification Pass Test
Additional Information Vapor Pressure: 29mbar at 20°C
Synonym Methylbenzene
MDL Number MFCD00008512
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Packaging Steel Pail
Chemical Name or Material Toluene
Cleanup Symbol Solvent
CAS 108-88-3
Health Hazard 1 Signal Word:DANGER!
Linear Formula C6H5CH3

Specifications

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade Laboratory Grade
Molecular Formula C7H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Synonym Methylbenzene
Accessories
 

Signal Word:DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

DANGER!

 

SDS & Safety Documents