missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene, 99.7% min, HPLC Grade, Alfa Aesar™

Manufacturer:  Alfa Aesar 22903K7

 View more versions of this product

Catalog No. AA22903K7



Specifications

Specifications

Toluene
C7H8
UN1294
14,9529
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
≥99.7%
0.867
1.4967
111°C
4L
108-88-3
MFCD00008512
635760
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CC1=CC=CC=C1
92.141
CHEBI:17578
Liquid
HPLC
Aromatic
4°C (40°F)
-93°C

Chemical Identifiers

108-88-3
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H315-H336;
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE

RTECSNumber : XS5250000

TSCA : TSCA

SDS Documents