Please login to your online account to display your discounted pricing

Toluene, HPLC Grade, 99.7% min, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade HPLC Grade
Molecular Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
UN Number UN1294
Beilstein 635760
Merck Index 14,9529
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA22903K7

  SDS
alfa aesar
22903K7
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade HPLC Grade
Formula Weight 92.14
Quantity 4L
Chemical Name or Material Toluene
TSCA TSCA
Odor Aromatic
Refractive Index 1.4967
Flash Point 4°C (40°F)
Boiling Point 111°C
Physical Form Liquid
Melting Point −93°C
Assay ≥99.7%
Molecular Formula C6H5CH3
MDL Number MFCD00008512
CAS 108-88-3
UN Number UN1294
Beilstein 635760
Merck Index 14,9529
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304-H361-H373-H315-H336;
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
Density 0.867
Health Hazard 3 GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE
RTECS Number XS5250000
Safety and Handling

Danger

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H361-H373-H315-H336;
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.