Please login to your online account to display your discounted pricing

Toluene (HPLC), Fisher Chemical


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  T290SK1

 View more versions of this product

Catalog No. T290SK-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Toluene
Cleanup Symbol Solvent
Name Note Also meets ACS specifications
Absorbance 1 AU max. at 286nm
CAS 108-88-3
ChemAlert Storage Symbol Red
Health Hazard 1 Signal Word:DANGER!
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
DOT Information DOT Class 3, : Flammable Liquid
Additional Information Vapor Pressure: 29mbar at 20°C
Purity Grade HPLC Grade
Purity Grade Notes Mobile phase for HPLC applications
Physical Form Liquid
Assay ≥99.8%
Assay Percent Range 99.8% min by GC
Molecular Formula C7H8
Formula Weight 92.14
MDL Number MFCD00008512
Synonym Methylbenzene
Density 0.866g/cm³
Identification Pass Test
Packaging Glass Bottle
Refractive Index 1.4930 to 1.4950 (at 25°C)
Residue after Evaporation 10ppm max.
Substances Darkened by H2SO4 Pass Test
Sulfur Compounds 0.003% max.
Vapor Pressure 29mbar at 20°C
Water 0.02% max.
Viscosity 0.6 mPa/s at 20°C
Boiling Point 111°C
Color Colorless
Melting Point −95°C
Quantity 1L

Specifications

Chemical Name or Material Toluene
Name Note Also meets ACS specifications
CAS 108-88-3
Purity Grade HPLC Grade
Purity Grade Notes Mobile phase for HPLC applications
Assay ≥99.8%
Assay Percent Range 99.8% min by GC
Molecular Formula C7H8
Formula Weight 92.14
MDL Number MFCD00008512
Synonym Methylbenzene
Accessories

Accessories for Catalog Number: T290SK1

Item Mfr. No. Item Description Price Per Unit Quantity  
296355104
Spilkleen/Cep  CEP LAB SPILL KIT SOLVENTS
spilkleen/cep
CSKLSK1SOL
Each for $341.99
Accessories
 

Signal Word:DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

DANGER!

 

SDS & Safety Documents