Please login to your online account to display your discounted pricing

Toluene (Histological), Fisher Chemical


Quantity 1 gal.
Packaging Poly Bottle

Manufacturer: fisher scientific company  BPT326F1GAL

 View more versions of this product

Catalog No. T326F-1GAL

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Purity Grade Histological Grade
Boiling Point 111°C
Melting Point −95°C
Formula Weight 92.14
Quantity 1 gal.
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Colorless
Density 0.866g/cm³
Vapor Pressure 29mbar at 20°C
Viscosity 0.6 mPa/s at 20°C
Synonym Methylbenzene
Health Hazard 1 Signal Word: Danger
Linear Formula C6H5CH3
Cleanup Symbol Solvent
Chemical Name or Material Toluene
CAS 108-88-3
MDL Number MFCD00008512
Packaging Poly Bottle
Additional Information Vapor Pressure: 29mbar at 20°C
Molecular Formula C7H8
Identification Pass Test
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health: 2
Flammability: 3
Instability: 0
Health Hazard 2 DANGER!
ChemAlert Storage Symbol Red

Specifications

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade Histological Grade
Assay ≥99.5%
Molecular Formula C7H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Synonym Methylbenzene
Accessories

Accessories for Catalog Number: T326F1GAL

Item Mfr. No. Item Description Price Per Unit Quantity  
296355104
Spilkleen/Cep  CEP LAB SPILL KIT SOLVENTS
spilkleen/cep
CSKLSK1SOL
Each for $341.99
Accessories
 

Signal Word: Danger

Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health: 2
Flammability: 3
Instability: 0

DANGER!