missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene (Histological), Fisher Chemical

Manufacturer:  Thermo Scientific BPT326S20

 View more versions of this product

Catalog No. T326S-20



Specifications

Specifications

Toluene
Vapor Pressure: 29mbar at 20°C
C6H5CH3
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CC1=CC=CC=C1
92.141
CHEBI:17578
Liquid
Histological
Pass Test
29mbar at 20°C
111°C
-95°C
108-88-3
C7H8
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
≥99.5%
0.866g/cm³
Steel Pail
0.6 mPa/s at 20°C
Colorless
20L

Chemical Identifiers

108-88-3
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health: 2
Flammability: 3
Instability: 0

Signal Word: Danger

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents