Please login to your online account to display your discounted pricing

Toluene For spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$204.97 - $204.97

Chemical Identifiers

Chemical Name or Material Toluene
CAS 108-88-3
Molecular Formula C7H8
Formula Weight 92.14
UN Number 1294
Synonym Toluol, Methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1083311000

 
emd millipore corporation
1083311000
Glass Bottle 1L Each for $204.97
Description & Specifications

Specifications

Formula Weight 92.14
Boiling Point 110.6°C
Melting Point −95°C
Quantity 1L
Description SupraSolv™ MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. . SupraSolv™ solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
Health Hazard 3 H315: Causes skin irritation.
Odor Charactristic
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.
Packaging Glass Bottle
Chemical Name or Material Toluene
Physical Form Liquid
Vapor Pressure 29 hPa (20°C)
Solubility Information Soluble in water: 0.52g/L at 16°C
Ignition Point 535°C (995°F)
Refractive Index 1.49
Flash Point 4.4°C (39.9°F), Closed cup
Molecular Formula C7H8
UN Number 1294
Synonym Toluol, Methylbenzene
Color Colorless
Density 0.87g/cm3 at 20°C
Health Hazard 2 H304: May be fatal if swallowed and enters airways.
CAS 108-88-3
Health Hazard 1 H225: Highly flammable liquid and vapour.
Safety and Handling

H225: Highly flammable liquid and vapour.

H315: Causes skin irritation.

H304: May be fatal if swallowed and enters airways.