Please login to your online account to display your discounted pricing

Toluene For gas chromatography ECD and FID SupraSolv™, EMD Millipore

For gas chromatography ECD and FID SupraSolv™

$149.77 - $314.00

Chemical Identifiers

Chemical Name or Material Toluene
CAS 108-88-3
Molecular Formula C7H8
Formula Weight 92.14
UN Number 1294
Synonym Methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1083891000

 
emd millipore corporation
1083891000
Glass Bottle 1L Each for $149.77

M1083892500

 
emd millipore corporation
1083892500
Glass Bottle 2.5L Each for $314.00
Description & Specifications

Specifications

Chemical Name or Material Toluene
CAS 108-88-3
Health Hazard 1 H225: Highly flammable liquid and vapour.
Health Hazard 2 H304: May be fatal if swallowed and enters airways.
Health Hazard 3 H315: Causes skin irritation.
Physical Form Liquid
Molecular Formula C7H8
Formula Weight 92.14
UN Number 1294
Synonym Methylbenzene
Solubility Information Soluble in water: 0.52g/L at 16°C
Description SupraSolv™ ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv™ solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
Density 0.87g/cm3 at 20°C
Ignition Point 535°C (995°F)
Odor Charactristic
Packaging Glass Bottle
Refractive Index 1.49
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.
Vapor Pressure 29 hPa (20°C)
Flash Point 4.4°C (39.9°F), Closed cup
Boiling Point 110.6°C
Color Colorless
Melting Point −95°C
Quantity 1L
Safety and Handling

H225: Highly flammable liquid and vapour.

H315: Causes skin irritation.

H304: May be fatal if swallowed and enters airways.