Please login to your online account to display your discounted pricing

Toluene, Environmental Grade, >99.8%, Alfa Aesar™

Suitable For Pesticide Residue Analysis

$102.01 - $102.01

Chemical Identifiers

Chemical Name or Material Toluene
Name Note Submicron filtered
CAS 108-88-3
Purity Grade Environmental Grade
Physical Form Liquid
Assay Percent Range >99.8%
Molecular Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43061K2

  SDS
alfa aesar
43061K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43061K7

  SDS
alfa aesar
43061K7
4L Each for $102.01

AA43061M6

  SDS
alfa aesar
43061M6
4 x 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Toluene
Name Note Submicron filtered
CAS 108-88-3
Health Hazard 1 H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.
DOT Information DOT Class: 3, Packaging Group: II
EINECS Number 203-625-9
Purity Grade Environmental Grade
Physical Form Liquid
Assay Percent Range >99.8%
Molecular Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Ignition Point 535°C (995°F)
Odor Aromatic
Vapor Pressure 22mm Hg at 20°C
Color Clear colorless
Quantity 1L

Maximum level of impurities: Color (APHA) 10, Halogenated residue by GC-ECD, (as Heptachlorepoxide) 5ng/L, Water 0.03%

Safety and Handling

H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.