Please login to your online account to display your discounted pricing

Toluene-d8, for NMR, 99.5+% atom D, ACROS Organics™

$30.83 - $145.08

Chemical Identifiers

Chemical Name or Material Toluene-d8
Name Note 99.5+ atom % D
CAS 2037-26-5
Purity Grade For NMR
Molecular Formula C7D8
Linear Formula C6D5CD3
Formula Weight 100.21
MDL Number MFCD00044638
Synonym Methylbenzene-d8
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC321320100

  SDS
thermo fisher scientific
321320100
Glass bottle 10mL Each for $145.08

AC321320010

  SDS
thermo fisher scientific
321320010
Glass bottle 1mL Each for $30.83
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 100.21
Quantity 10mL
Color Clear colorless
EINECS Number 218-009-5
Density 0.9400g/mL
Purity Grade For NMR
Beilstein 05, IV, 797
Chemical Name or Material Toluene-d8
Name Note 99.5+ atom % D
Molecular Formula C7D8
Linear Formula C6D5CD3
MDL Number MFCD00044638
CAS 2037-26-5
Synonym Methylbenzene-d8
Deuterium (D) 99.50% min.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

For NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.