Please login to your online account to display your discounted pricing

Toluene-d8, for NMR, 99.5% atom D, with 0.03% TMS, ACROS Organics™


Quantity 5mL

Manufacturer: thermo fisher scientific  351500050

 View more versions of this product

Catalog No. AC351500050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 100.21
Quantity 5mL
Color Clear colorless
Beilstein 05, IV, 797
Purity Grade For NMR
Density 0.9400g/mL
Deuterium (D) 99.4% min.
Chemical Name or Material Toluene-d8
CAS 2037-26-5
Name Note 99.5 atom%D
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym Methylbenzene-d8
Linear Formula C6D5CD3
MDL Number MFCD00044638
Molecular Formula C7D8

Specifications

Chemical Name or Material Toluene-d8
Name Note 99.5 atom%D
CAS 2037-26-5
Purity Grade For NMR
Molecular Formula C7D8
Linear Formula C6D5CD3
Formula Weight 100.21
MDL Number MFCD00044638
Synonym Methylbenzene-d8
Accessories

GHS Signal Word: Danger

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.

SDS & Safety Documents