Please login to your online account to display your discounted pricing

Alfa Aesar™ Toluene-d8, 100% (Isotopic)

Chemical Identifiers

Chemical Name or Material Toluene-d8
CAS 2037-26-5
Name Note Isotopic
Formula Weight 100.19
MDL Number MFCD00044638
UN Number UN1294
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42356KV

  SDS
alfa aesar™
42356KV
10 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42356SA

  SDS
alfa aesar™
42356SA
2 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.19
Packaging Prescored Ampules
Name Note Isotopic
Odor Aromatic
Melting Point −93°C
Density 0.94g/mL
Boiling Point 111°C
Assay 100%
Quantity 10 each
Health Hazard 1 Danger
MDL Number MFCD00044638
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE
Physical Form Liquid
Flash Point 10°C (50°F)
Health Hazard 3 GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
CAS 2037-26-5
UN Number UN1294
Chemical Name or Material Toluene-d8
Safety and Handling

Danger

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.