Please login to your online account to display your discounted pricing

Toluene, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Packaging Aluminum Bottle

Manufacturer: avantor performance  946005

 View more versions of this product

Catalog No. 02004470

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 92.14
Purity Grade Notes Meets ACS reagent chemical requirements
CAS 108-88-3
Chemical Name or Material Toluene
Packaging Aluminum Bottle
Specific Gravity 1L = 0.86kg
Flash Point 4.00°C
Quantity 4L
Molecular Formula C6H5CH3

Specifications

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade Baker Analyzed™ ACS Reagent
Purity Grade Notes Meets ACS reagent chemical requirements
Molecular Formula C6H5CH3
Formula Weight 92.14

    For Laboratory,Research,or Manufacturing Use