missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene, Anhydrous, 99.8% min., DriSolv, MilliporeSigma™

$113.97 - $179.71

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.141
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym antisal 1a
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MTX07327 View Documents Emd Millipore Corporation
TX07327
100mL Each for $113.97
 
MTX07326 View Documents Emd Millipore Corporation
TX07326
1L Each for $179.71
 
Specifications

Chemical Identifiers

108-88-3
92.141
antisal 1a
CHEBI:17578
CC1=CC=CC=C1
C7H8
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene

Specifications

108-88-3
C7H8
antisal 1a
CC1=CC=CC=C1
92.141
CHEBI:17578
Liquid
100mL
99.8%, min.
C6H5CH3
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
Undesignated
Toluene, Anhydrous, DriSolv™
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature