missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene, ACS Grade, ≥ 99.5 %, LabChem™

Supplier:  LabChem LC261704

 View more versions of this product

Catalog No. LC261704


Description

Description

Specifications

Specifications

Toluene
108-88-3
100
870kg/m3
4 L
Amber Glass
≤0.001%
C7H8
MFCD00008512
UN1294
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CC1=CC=CC=C1
92.14
CHEBI:17578
≥99.5%
-95°C
Colorless
Carbon dioxide; Carbon monoxide
111°C
Passes Test
Liquid
870kg/m3
C7H8
0.69mm2/s
≤0.03%
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
ACS

Chemical Identifiers

108-88-3
92.14
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
May cause damage

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at

Danger

EINECSNumber : 203-625-9

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions