Please login to your online account to display your discounted pricing

Toluene ACS, EMD Millipore


Quantity 20L
Packaging Metal Can

Manufacturer: emd millipore corporation  TX07353

 View more versions of this product

Catalog No. MTX07353

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 110.6°C
Assay ≥99.5%
Purity Grade ACS Reagent Grade
Melting Point −95°C
Formula Weight 92.14
Quantity 20L
Chemical Name or Material Toluene
Packaging Metal Can
Linear Formula C6H5CH3
CAS 108-88-3

Specifications

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade ACS Reagent Grade
Linear Formula C6H5CH3
Formula Weight 92.14