Please login to your online account to display your discounted pricing

Toluene ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade ACS Reagent Grade
Molecular Formula C6H5CH3
Formula Weight 92.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

14650374

 
avantor performance
860816
Glass Container 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Quantity 2.5L
Formula Weight 92.14
Molecular Formula C6H5CH3
CAS 108-88-3
Chemical Name or Material Toluene
Packaging Glass Container